Learn More
Tungsten wire, chromium plated, 1.8mm (0.07in) min dia, W 96.5+%, Cr 99.888% (metals basis), Thermo Scientific Chemicals
Evaporation source, contacts and coatings
Brand: Thermo Scientific Alfa Aesar 041040.8G
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Evaporation source, contacts and coatings
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tungsten wire, chromium plated | |
W ≥96.5%, Cr 99.888% (metals basis) | |
5 x 10 cm |
≤1.8mm (0.07 in.) dia., thick: 1.3mm (0.05 in.), Cr layer: ≤0.01 in. thick | |
MFCD00011461 | |
Wire |
RUO – Research Use Only