missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals

Catalog Number 11333548
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
62571-53-3
C12H30O6W
454.21
MFCD00156526
JHZFYDXXWXXVRM-UHFFFAOYSA-N
tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide
13828235
ethanolate;tungsten
CCO[W](OCC)(OCC)(OCC)(OCC)OCC
This item is not returnable. View return policy
62571-53-3
C12H30O6W
454.21
MFCD00156526
JHZFYDXXWXXVRM-UHFFFAOYSA-N
tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide
13828235
ethanolate;tungsten
CCO[W](OCC)(OCC)(OCC)(OCC)OCC

Tungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. It acts as an intermediate for sol-gel coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown to Yellow
Flash Point 17°C (63°F)
Odor Alcohol-like
Linear Formula W(OC2H5)6
Quantity 25 mL
UN Number UN1170
Sensitivity Moisture sensitive
Solubility Information Soluble in ethyl acetate,hydrocarbons and esters.
Formula Weight 454.21
Percent Purity 99.8%
Concentration or Composition (by Analyte or Components) 5% w/v in ethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tungsten(VI) ethoxide
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'