missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten(VI) ethoxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Intermediate for sol-gel coatingsTungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. Also, it acts as an intermediate for sol-gel coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 150°C |
Color | White to Brown |
Linear Formula | W(OC2H5)6 |
Quantity | 5 g |
UN Number | UN3181 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ethyl acetateSoluble in ethyl acetate,hydrocarbons and esters. |
Formula Weight | 454.21 |
Physical Form | Chunks and/or Powder |
Chemical Name or Material | Tungsten(VI) ethoxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'