missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(V) ethoxide 1,2-dimethoxyethane adduct, 99%, Thermo Scientific™

£47.40 - £147.00

Chemical Identifiers

Molecular Formula C10H25O5W·C4H10O2
MDL Number MFCD16294494
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15467045
View Documents Promotion Details
Thermo Scientific Alfa Aesar
045659.06
5 g
£47.40
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15477045
View Documents Promotion Details
Thermo Scientific Alfa Aesar
045659.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

C10H25O5W·C4H10O2
MFCD16294494

Specifications

<0°C
(decomposition)
W(OC2H5)5·C4H10O2
UN1993
Soluble in hydrocarbons. Reacts with water,alcohols,ketones and esters.
99%
Tungsten (V) ethoxide 1,2-dimethoxyethane adduct
Yellow
C10H25O5W·C4H10O2
MFCD16294494
Moisture sensitive
499.26
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H360-H332-H315-H319
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H332-H360FD

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications