missing translation for 'onlineSavingsMsg'
Learn More

Tungsten Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™

Product Code. 10680793 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
30 mL
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
30mL
500mL
This item is not returnable. View return policy

Product Code. 10680793

Brand: SPEX Certiprep PLW92X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
W1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-33-7, 7732-18-5
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS Min % 0.1, 99.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 99.9
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Water
Quantity 500 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
H2O    1000μg/mL    PLW9-2Y, -2X    10,000    PLW9-3Y, -3X
1% HNO3/2% HF    1000μg/mL    PLW2-2X
2% HNO3/5% HF    10,000    PLW2-3X
Chemical Name or Material Tungsten Standard
Show More Show Less

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.