missing translation for 'onlineSavingsMsg'
Learn More

Tungsten silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11377257
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377257 10 g 10g
11367257 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377257 Supplier Thermo Scientific Alfa Aesar Supplier No. 011551.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tungsten silicide is used in microelectronics as a contact material. It is also used as a shunt over polysilicon lines to increase their conductivity and increase signal speed. Further, it acts as a barrier layer between silicon and other metals. In addition to this, it is used in microelectro mechanical systems and for oxidation-resistant coatings. It is also employed as a replacement for earlier tungsten films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12039-88-2
Molecular Formula Si2W
Molecular Weight (g/mol) 240.01
MDL Number MFCD00049704
InChI Key WQJQOUPTWCFRMM-UHFFFAOYSA-N
Synonym tungsten disilicide, tungsten-silicide, tungsten silicide wsi2, tungsten silicide, w5si3
PubChem CID 16212546
IUPAC Name bis($l^{3}-silanylidyne)tungsten
SMILES [Si]#[W]#[Si]

Specifications

Melting Point 2165°C
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 240.02
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Tungsten silicide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.