missing translation for 'onlineSavingsMsg'
Learn More

Tungsten rod, 6.3mm (0.25in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11335898
Click to view available options
Quantity:
10 cm
50 cm
This item is not returnable. View return policy
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
23964
CHEBI:27998
tungsten
[W]
This item is not returnable. View return policy
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
23964
CHEBI:27998
tungsten
[W]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance Silver Gray rod
Assay Percent Range 99.95% (Metals basis)
Quantity 10 cm
Form Rod
Chemical Name or Material Tungsten

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'