missing translation for 'onlineSavingsMsg'
Learn More

Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL, Thermo Scientific Chemicals

£55.70 - £300.00

Chemical Identifiers

Molecular Formula W in 5% HNO3/tr. HF
MDL Number MFCD00011461
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388667
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035772.AD
50 mL
£55.70
50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11378667
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035772.AE
100 mL
£100.00
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11485070
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035772.AP
500 mL
£300.00
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

W in 5% HNO3/tr. HF
MFCD00011461

Specifications

W 10,000μg/ml
MFCD00011461
Acidic
Tungsten, plasma standard solution, Specpure™
W in 5% HNO3/tr. HF
UN3264
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H332;
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only