missing translation for 'onlineSavingsMsg'
Learn More

Tungsten hexacarbonyl, 97%

Product Code. 10062973 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10062973

Brand: Thermo Scientific Acros 221042500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tungsten hexacarbonyl, 99%, C6O6W, CAS Number-14040-11-0, 250g, 175.0 deg.C, Yellow to Beige, 237-880-2, 98.5% min., 06,658; 08,525; 05,142; 06,125; 07,71; 08,110; 13,51, 351.91, GHS Signal Word: Danger, GHS H Statement Toxic in contact with skin. Toxic if swallowed. Toxic if inhaled. | CAS: 14040-11-0 | C6O6W | 351.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14040-11-0
Molecular Formula C6O6W
Molecular Weight (g/mol) 351.91
MDL Number MFCD00011462

Specifications

Boiling Point 175.0°C
Melting Point 150.0°C
Color Yellow to Beige
Physical Form Fine Crystalline Powder
Quantity 250 g
Assay Percent Range 99%
Linear Formula W(CO6)
Fieser 06,658; 08,525; 05,142; 06,125; 07,71; 08,110; 13,51
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 351.91
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Tungsten hexacarbonyl
Show More Show Less
compliance-icons
Product Identifier
  • Tungsten hexacarbonyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.