missing translation for 'onlineSavingsMsg'
Learn More

Tungsten gauze, gold plated, 50 mesh woven from 0.025mm (0.001in) dia wire

Product Code. 11395227
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395227 300 x 300 mm Each
11375227 75 x 75 mm Each
11385227 150 x 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11395227 Supplier Thermo Scientific Alfa Aesar Supplier No. 040926.HB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Form: Wire Cloth; Open area: 90.3%; Width of opening: 0.019 in. | CAS: 7440-33-7 | W | 183.84 g/mol

Filtration, electrodesTungsten gauze, gold plated, wire is used in filtration. It also serves as an electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 300 x 300 mm
Mesh Size 50 Mesh
Form Wire Cloth
Solubility Information Soluble in mixture of nitric acid and hydrofluoric acid. Insoluble in water.
Chemical Name or Material Tungsten

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.