missing translation for 'onlineSavingsMsg'
Learn More

Tungsten Gauze, 20 mesh woven from 0.127mm (0.005 in.) dia. .wire, Alfa Aesar™

Form: Wire Cloth; Open area: 81%; Width of opening: 0.045 in.

Brand:  Alfa Aesar 040922.FL

Code : NEW

Additional Details : CAS Number : 7440-33-7 Weight : 0.01000kg

Quantity 75 x 75mm

 View more versions of this product

Product Code. 11355217

  • £114.80 / Each


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Tungsten
7440-33-7
Gauze
MFCD00011461

Specifications

Tungsten
7440-33-7
MFCD00011461
75 x 75mm
Gauze
Videos
SDS