missing translation for 'onlineSavingsMsg'
Learn More

Tungsten dichloride dioxide, 99%

Product Code. 11483870
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483870 10 g 10g
11473870 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483870 Supplier Thermo Scientific Alfa Aesar Supplier No. 089962.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13520-76-8 | WO2Cl2

Tungsten dichloride dioxide is used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13520-76-8
Molecular Formula WO2Cl2
MDL Number MFCD00054136

Specifications

Color Yellow
Physical Form Powder
Quantity 10 g
Assay Percent Range 99%
Solubility Information Soluble in cold water. Decomposes in hot water.
Formula Weight 286.75
Odor Acrid
Sensitivity Moisture sensitive
Chemical Name or Material Tungsten dichloride dioxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.