missing translation for 'onlineSavingsMsg'
Learn More

Tungsten carbide Cobalt, 99% (metals basis)

Product Code. p-7049803
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368227 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11368227 Supplier Thermo Scientific Alfa Aesar Supplier No. 022955.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12774-15-1 | CHCoW | 255.79 g/mol

Tungsten carbide Cobalt is used in cutting tools, having hard ceramic crystals of a few microns in size within a ductile metal matrix, use as electrodes for Electrode Discharge Machining. It is also used in production of tools for milling, drilling and pressing/punching processes, surgical instruments, product is the rotating balls used in ball point pens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12774-15-1
Molecular Formula CHCoW
Molecular Weight (g/mol) 255.79
MDL Number MFCD00064847
InChI Key IKEHVRMRUVYPGG-UHFFFAOYSA-N
Synonym tungsten carbide cobalt, methylidynetungsten cobalt
PubChem CID 56846058
SMILES [Co].C#[W]

Specifications

Color Gray
Avg. Mol. Wt. or Mol. Wt. Range WC:Co; 94:6 wt%
Quantity 1 kg
Mesh Size −325 Mesh
Odor Odorless
Form Powder
UN Number UN3178
Solubility Information Insoluble in water.
IUPAC Name methylidynetungsten cobalt
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Tungsten carbide Cobalt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.