missing translation for 'onlineSavingsMsg'
Learn More

Tungsten carbide, 99.5% (metals basis)

Product Code. p-4904562
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11301829

Brand: Thermo Scientific Alfa Aesar 012482.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12070-12-1 | CW | 195.85 g/mol

Tungsten carbide is extensively used as a cutting tool for machining and in armor-piercing ammunition. It is also used in farriery, the shoeing of horses as well as to improve traction on slippery surfaces like roads or ice. It finds application in nuclear reactors as an effective neutron reflector. It is also used for making surgical instruments meant for open surgery such as scissors, forceps, hemostats, blade-handles and in laparoscopic surgery instruments such as graspers, cutter, needle holder and cautery. Further, it also used to make the rotating ball in the tips of ballpoint pens, bridal jewelry, and gauge blocks, which is useful for producing precision lengths in dimensional metrology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-12-1
Molecular Formula CW
Molecular Weight (g/mol) 195.85
MDL Number MFCD00011464
InChI Key UONOETXJSWQNOL-UHFFFAOYSA-N
Synonym tungsten carbide, tungsten iv carbide, rfdbds g@, tungsten carbide ds 60, methanidylidynetungstenylium, methanidylidynetungsten 1+, tungsten-carbide/cobalt co=8wt% powder nano
IUPAC Name methanidylidynetungstenylium
SMILES [C-]#[W+]

Specifications

Melting Point 2870°C
Color Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5% metal basis
Solubility Information Soluble in nitric acid and hydrofluroic acid. Insoluble in water.
Formula Weight 195.86
Odor Odorless
Density 15.63
Chemical Name or Material Tungsten carbide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.