missing translation for 'onlineSavingsMsg'
Learn More

Tungsten carbide, 99.5% (metals basis)

Código de producto. 11391819
Change view
Click to view available options
Quantity:
100 g
500 g
Tamaño de la unidad:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
11391819 500 g 500g
11301829 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11391819 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 012482.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 12070-12-1 | CW | 195.85 g/mol

Tungsten carbide is extensively used as a cutting tool for machining and in armor-piercing ammunition. It is also used in farriery, the shoeing of horses as well as to improve traction on slippery surfaces like roads or ice. It finds application in nuclear reactors as an effective neutron reflector. It is also used for making surgical instruments meant for open surgery such as scissors, forceps, hemostats, blade-handles and in laparoscopic surgery instruments such as graspers, cutter, needle holder and cautery. Further, it also used to make the rotating ball in the tips of ballpoint pens, bridal jewelry, and gauge blocks, which is useful for producing precision lengths in dimensional metrology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 12070-12-1
Molecular Formula CW
Molecular Weight (g/mol) 195.85
MDL Number MFCD00011464
InChI Key UONOETXJSWQNOL-UHFFFAOYSA-N
Synonym tungsten carbide, tungsten iv carbide, rfdbds g@, tungsten carbide ds 60, methanidylidynetungstenylium, methanidylidynetungsten 1+, tungsten-carbide/cobalt co=8wt% powder nano
IUPAC Name methanidylidynetungstenylium
SMILES [C-]#[W+]

Especificaciones

Melting Point 2870°C
Color Gray
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5% metal basis
Solubility Information Soluble in nitric acid and hydrofluroic acid. Insoluble in water.
Formula Weight 195.86
Odor Odorless
Density 15.63
Chemical Name or Material Tungsten carbide

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.