missing translation for 'onlineSavingsMsg'
Learn More

Tungsten carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11388808
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
12070-13-2
CW2
379.69
MFCD00801036
FHLFYDOXSZQEPO-UHFFFAOYSA-N
tungsten carbide w2c, methyliumylidyneditungstenene
102601578
methyliumylidyneditungstenene
[W-]=[W]#[C+]
This item is not returnable. View return policy
12070-13-2
CW2
379.69
MFCD00801036
FHLFYDOXSZQEPO-UHFFFAOYSA-N
tungsten carbide w2c, methyliumylidyneditungstenene
102601578
methyliumylidyneditungstenene
[W-]=[W]#[C+]

Tungsten carbide is used in industrial machinery, cutting tools, abrasives, armor-piercing rounds and jewelry. It is preferred even over stainless steel because of their incredible hardness and resistance to wear and tear. It is utilized for the manufacture of mill products including grinding and milling. It also includes tipping trekking, ski poles as well as cleats. It is mainly used in the form of cemented tungsten carbide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼2800°C
Color Black
Physical Form Powder
Quantity 50 g
Linear Formula W2C
UN Number UN3178
Solubility Information Soluble in nitric acid and hydrofluoric acid. Insoluble in water.
Formula Weight 379.71
Percent Purity 99.5%
Density 17.15 g/mL
Assay (metals basis)
Chemical Name or Material Tungsten carbide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'