missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tumor Necrosis Factor-alpha, human

Product Code. p-7076719
Click to view available options
Quantity:
10μg
50μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 15428659

Brand: Thermo Scientific Chemicals J67162.I&

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 17.5 kd
Quantity 50μg
Packaging Plastic bottle
Form Lyophilized

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.