missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TULP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92546
This item is not returnable.
View return policy
Description
TULP4 Polyclonal specifically detects TULP4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TULP4 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 | |
| KIAA1397tubby super-family protein, tubby like protein 4, Tubby superfamily protein, Tubby-like protein 4, TUBL4, TUSPtubby-related protein 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56995 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TULP4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VLLHESDGVLGMSWNYPIFLVEDSSESDTDSDDYAPPQDGPAAYPIPVQNIKPLLTVSFTSGDISLMNNYDDLSPTVIRSGLKEVVAQWCTQGDLLAVAGMERQTQLGELPNGPLLKSAMVKFYNVRGEHIFTLDTLVQRPIISICWGH | |
| 0.1 mL | |
| Primary | |
| Specificity of human TULP4 antibody verified on a Protein Array containing target protein plus 96 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction