missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TULP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553297
This item is not returnable.
View return policy
Description
Immunogen sequence: TGTSGSATAA QPADNLLGDI DDLEDFVYSP APQGVTVRCR IIRDKRGMDR GLFPTYYMYL EKEENQKIFL LAARKRKKSK TANYLISIDP VDLSREGESY VGK Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 72%.
This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.
Specifications
| TULP3 | |
| Polyclonal | |
| Unconjugated | |
| TULP3 | |
| 2310022L06Rik; AI316887; TUB like protein 3; tubby like protein 3; tubby-like protein 3; Tubby-related protein 3; TUBL3; Tulp3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7289 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75386 | |
| TULP3 | |
| Recombinant protein corresponding to Human TULP3. Recombinant protein control fragment (Product #RP-91604). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction