missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TULP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13637-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.Specifications
| TULP3 | |
| Polyclonal | |
| Unconjugated | |
| TULP3 | |
| tubby like protein 3, Tubby related protein 3, TUBL3, TULP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22158, 688749, 7289 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75386, O88413 | |
| TULP3 | |
| TULP3 Fusion Protein Ag4570 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction