missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TULP3 Monoclonal Antibody (OTI4E5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525831
This item is not returnable.
View return policy
Description
TULP3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.
Specifications
| TULP3 | |
| Monoclonal | |
| 0.78 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75386 | |
| TULP3 | |
| Full length human recombinant protein of TULP3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI4E5 | |
| Unconjugated | |
| TULP3 | |
| 2310022L06Rik; AI316887; TUB like protein 3; tubby like protein 3; tubby-like protein 3; Tubby-related protein 3; TUBL3; Tulp3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7289 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction