missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TULP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | TULP2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TULP2 Polyclonal specifically detects TULP2 in Human samples. It is validated for Western Blot.Specifications
| TULP2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS buffer, 2% sucrose | |
| 7288 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Cancer/testis antigen 65, CT65cancer testis antigen 65, tubby like protein 2, Tubby-like protein 2, TUBL2tubby-related protein 2 | |
| The immunogen is a synthetic peptide directed towards the middle region of human TULP2 (NP_003314). Peptide sequence RKRRRSKTSNYLISLDPTLLSRDGDNFVGKVRSNVFSTKFTIFDNGVNPD | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title