missing translation for 'onlineSavingsMsg'
Learn More

Tulobuterol hydrochloride, MedChemExpress

Product Code. 16452506
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452506 500 mg 500mg
16419356 100 mg 100mg
16405457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16452506 Supplier Medchem Express Supplier No. HYW011733
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tulobuterol hydrochloride (C-78) is a long-acting β2-adrenoceptor agonist, which reduces the frequency of exacerbations of chronic obstructive pulmonary disease and bronchial asthma. Tulobuterol hydrochloride is also a sympathomimetic agent used as a transdermal patch, increases normal diaphragm muscle strength. Tulobuterol hydrochloride inhibit rhinovirus replication and modulate airway inflammation.

Tulobuterol hydrochloride (C-78) is a long-acting β2-adrenoceptor agonist, which reduces the frequency of exacerbations of chronic obstructive pulmonary disease and bronchial asthma. Tulobuterol hydrochloride is also a sympathomimetic agent used as a transdermal patch, increases normal diaphragm muscle strength. Tulobuterol hydrochloride inhibit rhinovirus replication and modulate airway inflammation.

Specifications

Chemical Name or Material Tulobuterol hydrochloride
Physical Form Solid
CAS 56776-01-3
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C12H19Cl2NO
For Use With (Application) COVID-19-anti-virus
Synonym C-78
Solubility Information H2O : 100 mg/mL (378.52 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (189.26 mM; Need ultrasonic)
SMILES CC(C)(NCC(O)C1=CC=CC=C1Cl)C.Cl
Molecular Weight (g/mol) 264.19
Formula Weight 264.19
Percent Purity 99.69%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.