missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tulisokibart Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA555016

Product Code. 30131927

  • £387.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Compatible with Direct ELISA.

Tulisokibart is under development for the treatment of inflammatory bowel diseases like ulcerative colitis, crohn's disease and systemic sclerosis-associated interstitial lung disease (SSc-ILD). The drug comprises of humanized IgG1 monoclonal antibody and is administered through intravenous and subcutaneous route.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tulisokibart Humanized
Recombinant Monoclonal
Unconjugated
MK-7240; PRA-023
Recombinant Human TNFSF15/TL1A.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
4.39 mg/mL
PBS with no preservative
Human
Protein A/G
RUO
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.