missing translation for 'onlineSavingsMsg'
Learn More

Tulathromycin A, TRC

Product Code. 17964225
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964225 50 mg 50mg
17913105 5 mg 5mg
17981595 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964225 Supplier Toronto Research Chemicals Supplier No. TRCT89715050MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 217500-96-4
Molecular Formula C41 H79 N3 O12
Molecular Weight (g/mol) 806.08
Synonym (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-13-[[2,6-Dideoxy-3-C-methyl-3-O-methyl-4-C-[(propylamino)methyl]-α-L-ribo-hexopyranosyl]oxy]-2-ethyl-3,4,10-trihydroxy-3,5,8,10,12,14-hexamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-1-oxa-6-azacyclopentadecan-15-one, CP 472295, Draxxin, Tulathromycin, Tulathromycin A
IUPAC Name (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyl-5-(propylaminomethyl)oxan-2-yl]oxy-3,5,8,10,12,14-hexamethyl-1-oxa-6-azacyclopentadecan-15-one
SMILES CCCNC[C@]1(O)[C@H](C)O[C@H](C[C@@]1(C)OC)O[C@H]2[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN[C@H](C)[C@@H](O)[C@](C)(O)[C@@H](CC)OC(=O)[C@@H]2C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C41H79N3O12/c1-15-17-42-22-41(50)28(8)53-31(20-39(41,10)51-14)55-33-25(5)35(56-37-32(45)29(44(12)13)18-24(4)52-37)38(9,48)19-23(3)21-43-27(7)34(46)40(11,49)30(16-2)54-36(47)26(33)6/h23-35,37,42-43,45-46,48-50H,15-22H2,1-14H3/t23-,24-,25+,26-,27-,28+,29+,30-,31+,32-,33+,34-,35-,37+,38-,39-,40-,41+/m1/s1
Formula Weight 805.5664
Percent Purity >95%
Chemical Name or Material Tulathromycin A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.