missing translation for 'onlineSavingsMsg'
Learn More

Tulathromycin A, MedChemExpress

Product Code. 16492807
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492807 50 mg 50mg
16476866 100 mg 100mg
16476728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492807 Supplier Medchem Express Supplier No. HY15662
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tulathromycin A (Tulathromycin), a macrolide antibiotic, inhibits protein synthesis (IC50=0.26 μM) by targeting bacterial ribosome. Tulathromycin A is used for the research of respiratory disease in cattle and swine. Immunomodulatory effects.

Tulathromycin A (Tulathromycin), a macrolide antibiotic, inhibits protein synthesis (IC50=0.26 μM) by targeting bacterial ribosome. Tulathromycin A is used for the research of respiratory disease in cattle and swine. Immunomodulatory effects.

Specifications

Chemical Name or Material Tulathromycin A
Physical Form Solid
CAS 217500-96-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C41H79N3O12
For Use With (Application) COVID-19-immunoregulation
Synonym Tulathromycin CP 472295
Solubility Information DMSO : 50 mg/mL (62.03 mM; Need ultrasonic)
SMILES O=C1O[C@H](CC)[C@@](C)(O)[C@H](O)[C@@H](C)NC[C@H](C)C[C@@](C)(O)[C@H](O[C@@]2([H])[C@H](O)[C@@H](N(C)C)C[C@@H](C)O2)[C@@H](C)[C@H](O[C@@]3([H])C[C@@](C)(OC)[C@@](CNCCC)(O)[C@H](C)O3)[C@H]1C
Molecular Weight (g/mol) 806.08
Formula Weight 806.08
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.