missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TULA/STS-2 Monoclonal antibody specifically detects TULA/STS-2 in Human samples. It is validated for ELISA, ELISA
Specifications
Specifications
| Antigen | TULA/STS-2 |
| Applications | ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 1G6 |
| Conjugate | Unconjugated |
| Dilution | ELISA, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_001001895 |
| Gene Alias | Cbl-interacting protein 4, gene similar to UBA containing SH3 domain10CLIP4STS-2T-cell ubiquitin ligand protein, STS2, Suppressor of T-cell receptor signaling 2, T-cell ubiquitin ligand, TULAubiquitin-associated and SH3 domain-containing protein A, ubiquitin associated and SH3 domain containing A |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?