missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TULA/STS-2 Antibody (101), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90242AF532
This item is not returnable.
View return policy
Description
TULA/STS-2 Monoclonal specifically detects TULA/STS-2 in Human samples. It is validated for ELISA.
Specifications
| TULA/STS-2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 53347 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 101 | |
| ELISA | |
| Cbl-interacting protein 4, gene similar to UBA containing SH3 domain10CLIP4STS-2T-cell ubiquitin ligand protein, STS2, Suppressor of T-cell receptor signaling 2, T-cell ubiquitin ligand, TULAubiquitin-associated and SH3 domain-containing protein A, ubiquitin associated and SH3 domain containing A | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human TULA/STS-2 (AAH69511.1; Ala354-Asn623). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction