missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tug Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113355
This item is not returnable.
View return policy
Beskrivning
Tug Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene is a candidate gene for alveolar soft part sarcoma (ASPS). It has been found that this gene is fused with transcription factor TFE3 gene in ASPS and also in renal cell carcinomas. Several alternatively spliced transcript variants of this gene have been described, but their full length nature has not been determined.
Specifikationer
| Tug | |
| Polyclonal | |
| Unconjugated | |
| ASPSCR1 | |
| 1190006K01Rik; alveolar soft part sarcoma chromosomal region candidate gene 1 protein; Alveolar soft part sarcoma chromosomal region candidate gene 1 protein homolog; alveolar soft part sarcoma chromosome region, candidate 1; alveolar soft part sarcoma chromosome region, candidate 1 (human); alveolar soft part sarcoma locus; ASPC; ASPCR1; ASPL; ASPS; Aspscr1; ASPSCR1, UBX domain containing tether for SLC2A4; RCC17; renal cell carcinoma gene on chromosome 17; renal cell carcinoma, papillary, 17; renal papillary cell carcinoma protein 17; Tether containing UBX domain for GLUT4; tether, containing a UBX domain, for GLUT4; TUG; UBX domain protein 9; UBX domain-containing protein 9; UBXD9; UBXN9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79058 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.78 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BZE9 | |
| ASPSCR1 | |
| Recombinant Human Tether containing UBX domain for GLUT4 protein (284-553AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering