missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUG Antibody (SR2139), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22469-50ul
This item is not returnable.
View return policy
Description
TUG Monoclonal antibody specifically detects TUG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| TUG | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR2139 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Alveolar soft part sarcoma chromosomal region candidate gene 1 protein, alveolar soft part sarcoma chromosome region, candidate 1, Alveolar soft part sarcoma locus, ASPLUBX domain-containing protein 9, ASPSrenal cell carcinoma gene on chromosome 17, RCC17renal cell carcinoma, papillary, 17, Renal papillary cell carcinoma protein 17, tether containing UBX domain for GLUT4, TUG, UBX domain protein 9, UBXD9ASPCR1, UBXN9FLJ45380 | |
| A synthesized peptide derived from human TUG (Uniprot #: Q9BZE9) | |
| 50 μg | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 79058 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction