missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TUG Polyclonal antibody specifically detects TUG in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TUG |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Alveolar soft part sarcoma chromosomal region candidate gene 1 protein, alveolar soft part sarcoma chromosome region, candidate 1, Alveolar soft part sarcoma locus, ASPLUBX domain-containing protein 9, ASPSrenal cell carcinoma gene on chromosome 17, RCC17renal cell carcinoma, papillary, 17, Renal papillary cell carcinoma protein 17, tether containing UBX domain for GLUT4, TUG, UBX domain protein 9, UBXD9ASPCR1, UBXN9FLJ45380 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 503 and 553 of human alveolar soft part sarcoma locus using the numbering given in entry NP_076988.1 (GeneID 79058). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?