missing translation for 'onlineSavingsMsg'
Learn More

TUG-770, MedChemExpress

Product Code. 16489297
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489297 5 mg 5mg
16466748 10 mg 10mg
16462827 50 mg 50mg
16416886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489297 Supplier Medchem Express Supplier No. HY15697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TUG-770 is a potent, selective and orally active GPR40/FFA1 agonist with an EC50 of 6 nM for human FFA1. TUG-770 shows a high selectivity for FFA1 over FFA2, FFA3, FFA4, PPARγ, other receptors, transporters, and enzymes. TUG-770 can be uesd for type 2 diabetes research.

TUG-770 is a potent, selective and orally active GPR40/FFA1 agonist with an EC50 of 6 nM for human FFA1. TUG-770 shows a high selectivity for FFA1 over FFA2, FFA3, FFA4, PPARγ, other receptors, transporters, and enzymes. TUG-770 can be uesd for type 2 diabetes research.

Specifications

Chemical Name or Material TUG-770
CAS 1402601-82-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H14FNO2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (325.39 mM)
SMILES FC1=C(CCC(O)=O)C=CC(C#CC2=C(CC#N)C=CC=C2)=C1
Molecular Weight (g/mol) 307.32
Formula Weight 307.32
Percent Purity 99.59%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.