missing translation for 'onlineSavingsMsg'
Learn More

HC Innovations TuffScrub Two Sided Heavy Duty Wet Wipe

Product Code. 17274204
Click to view available options
Dimensions (L x W):
200 x 200 mm
This item is not returnable. View return policy

Product Code. 17274204

Brand: HC Innovations CP150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This 2 sided extremely tough wipe is formulated for removing grime, grease, oils, paints, ink, silicone and adhesives

  • Can be used on hands and surfaces.
  • This wipe comes in a heavy duty dispensing canister that holds 80 Large 2 sided wipes.
  • One side is textured giving you that little bit more for getting off anything that's stuck on.
  • Side 2 is soft for doing a second wipe down just to remove the last bit of dirt or grime.
  • These heavy duty wipes can be used on glass, windows, doors and lights

Specifications

Product Type Sanitizing Wipe
Material Tuf-Scrub
Autoclavable Not Autoclavable
Basis Weight 60 gsm
Color Turquoise
Content And Storage Store in a Dry Place
Disposable Single-use
Edges Angled
Dimensions (L x W) 200 x 200 mm
For Use With (Application) Removing Grime, Grease, Oils
For Use With (Equipment) Machinery
Packaging Quantity 80 Wipes
Ply 1-ply
Saturation Solution 10-30% Ethanol up to 1 % Dipentene
Sterility Non-sterile
Includes Fabric, Chemical
Green Features Recyclable
Care Instructions Keep upright
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.