missing translation for 'onlineSavingsMsg'
Learn More

Tubulin Beta 4 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18333744 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333744 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333744 Supplier Novus Biologicals Supplier No. NBP299382UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Tubulin Beta 4 Polyclonal specifically detects Tubulin Beta 4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin Beta 4
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias TUBB5beta-5, Tubulin 5 beta, tubulin beta-4 chain, tubulin, beta 4, tubulin, beta, 5
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Tubulin Beta 4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 10382
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.