missing translation for 'onlineSavingsMsg'
Learn More

Tubulin alpha-8 Antibody (OTI2G6), DyLight 594, Novus Biologicals™

Product Code. 18411365 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411365 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411365 Supplier Novus Biologicals Supplier No. NBP274709DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tubulin alpha-8 Monoclonal antibody specifically detects Tubulin alpha-8 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin alpha-8
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2G6
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Alpha-tubulin 8, TUBAL2Tubulin alpha chain-like 2, tubulin alpha-8 chain, tubulin, alpha 8
Host Species Mouse
Immunogen Full length recombinant protein of human Tubulin alpha-8 (NP_061816) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51807
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.