missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin alpha-1B, ac Lys40 Antibody (RM318), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-61591
This item is not returnable.
View return policy
Description
Tubulin alpha-1B Monoclonal antibody specifically detects Tubulin alpha-1B in Vertebrate samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Tubulin alpha-1B | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Vertebrates | |
| Purified |
| Western Blot, Immunofluorescence | |
| RM318 | |
| Western Blot 1:1000 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:1000-1:5000 | |
| alpha tubulin, Alpha-tubulin ubiquitous, K-ALPHA-1, tubulin alpha-1B chain, Tubulin alpha-ubiquitous chain, Tubulin K-alpha-1, tubulin, alpha 1b, tubulin, alpha, ubiquitous | |
| An acetyl-peptide corresponding to Acetyl-Tubulin alpha-1B [ac Lys40] | |
| 100 μg | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| 10376 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction