Learn More
Restek Tubing Sulfinert .180"ID x 1/4"OD Sulfinert Seamless 316L Grade 0.035" Thick Wall 100ft Roll
Brand:  Restek 29081
Encompass_Preferred
Tubing Sulfinert .180"ID x 1/4"OD Sulfinert Seamless 316L Grade 0.035" Thick Wall 100ft Roll
Product Code. 16648886
  • £4228.95 / 1 roll