Learn More
Upchurch Scientific Tubing SST yellow 1/16 inch O.D., 0.010 inch(0.25mm) I.D.1.52m
Brand:  Upchurch Scientific 02010-86
Tubing SST yellow 1/16 inch O.D., 0.010 inch(0.25mm) I.D.1.52m
Product Code. 13040850
  • £55.25 / Each
Only null left
Estimated Shipment: 26-01-2026