Learn More
Upchurch Scientific Tubing sleeve PEEK(TM) blue 13 inch (33mm) 1/16inch O.D., 865µm I.D. for 785 to 825µm O.D. tubing
Brand:  Upchurch Scientific 02017-94
Tubing sleeve PEEK(TM) blue 13 inch (33mm) 1/16inch O.D., 865µm I.D. for 785 to 825µm O.D. tubing
Product Code. 13083090
  • £7.40 / Each
Only null left
Estimated Shipment: 28-01-2026