missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Tubing Sinker

Product Code. 12992570 Shop All SP Scienceware Products
Click to view available options
For Use With:
5.5 to 6.2mm I.D. tubing
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12992570

Brand: Bel-Art™ SP Scienceware™ F329030102

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for use with pumps and dispensers

  • Teflon™ sinkers keep tubing at the bottom of your container
  • Hose barb ends for simple tubing connection
  • Solvent resistant and autoclavable

Specifications

Autoclavable Autoclavable
Material Teflon
For Use With 5.5 to 6.2mm I.D. tubing
Color White
Flexibility Rigid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.