missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interscience Silicone Tubing with Filter
Description
Silicone tubing with filter for cleaning pump
Specifications
Specifications
| Description | Sillicone tubing with filter |
| Autoclavable | Not Autoclavable |
| Material | Silicone |
| For Use With | easySpiral |
| For Use With (Application) | Silicone tubing with filter for cleaning pump |
| Packaging Quantity | 1 |
| Sterility | Non-sterile |
| Color | Clear |
| Product Type | Tubing |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?