Learn More
Smiths Medical Tubing Portex Non-Toxic
Product Code. 13160873
Encompass
Product Code. 13160873 Supplier Smiths Medical Supplier No. 800/500/525
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
non-contaminating silicone rubber for use with peristaltic pumps, 5m roll translucent -17 to +250 degrees ID 10mm x OD 15mm Smiths Medical