Learn More
Masterflex™ Tubing Adapter, PVDF Female Luer Lock
Click to view available options
Material:
PVDF
Unit Size:
Each
Description
These Female luer lock x 3/4 in. mini sanitary adapters are available in either polypropylene or PVDF.
Specifications
Specifications
| Description | Sanitary Tubing Adapter, PVDF; 3/4 in. Mini to Female Luer Lock |
| Material | PVDF |
| Outlet Port | Sanitary x Luer |
| Includes | Sanitary Clamp Size (Port 1): 3/4 in. |
| Material (Seal) | PVDF |
| Product Type | Vacuum Tubing |