missing translation for 'onlineSavingsMsg'
Learn More

TUBGCP5 Antibody (2H5), Novus Biologicals™

Product Code. 18401279 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401279 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401279 Supplier Novus Biologicals Supplier No. H00114791M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TUBGCP5 Monoclonal antibody specifically detects TUBGCP5 in Human samples. It is validated for ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBGCP5
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2H5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_443135
Gene Alias gamma-tubulin complex component 5, gamma-tubulin complex component GCP5, GCP5KIAA1899GCP-5, tubulin, gamma complex associated protein 5
Host Species Mouse
Immunogen TUBGCP5 (NP_443135.2, 1 a.a. ∽ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MARHGPPWSRLDAQQERDVRELVRGVAGLQDEADPNFQLALNFAWSNFRFHRFLDVNSHKIEKTIEGIYEKFVIHSDLSKAASWKRLTEEFLNAPLPSI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 114791
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.