missing translation for 'onlineSavingsMsg'
Learn More

TUBGCP3 Antibody, Novus Biologicals™

Product Code. 18329488 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329488 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329488 Supplier Novus Biologicals Supplier No. H00010426B03P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TUBGCP3 Polyclonal antibody specifically detects TUBGCP3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBGCP3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_006313.1
Gene Alias Gamma-ring complex protein 104 kDa, gamma-tubulin complex component 3, GCP-3, GCP3h104p, hGCP3, hGrip104, hSpc98, SPBC98, Spc98p, Spindle pole body protein Spc98 homolog, tubulin, gamma complex associated protein 3
Host Species Mouse
Immunogen TUBGCP3 (NP_006313.1, 1 a.a. - 907 a.a.) full-length human protein. MATPDQKSPNVLLQNLCCRILGRSEADVAQQFQYAVRVIGSNFAPTVERDEFLVAEKIKKELIRQRREADAALFSELHRKLHSQGVLKNKWSILYLLLSLSEDPRRQPSKVSSYATLFAQALPRDAHSTPYYYARPQTLPLSYQDRSAQSAQSSGSVGSSGISSIGLCALSGPAPAPQSLLPGQSNQAPGVGDCLRQQLGSRLAWTLTANQPSSQATTSKGVPSAVSRNMTRSRREGDTGGTMEITEAALVRDILYVFQGIDGKNIKMNNTENCYKVEGKANLSRSLRDTAVRLSELGWLHNKIRRYTDQRSLDRSFGLVGQSFCAALHQELREYYRLLSVLHSQLQLEDDQGVNLGLESSLTLRRLLVWTYDPKIRLKTLAALVDHCQGRKGGELASAVHAYTKTGDPYMRSLVQHILSLVSHPVLSFLYRWIYDGELEDTYHEFFVASDPTVKTDRLWHDKYTLRKSMIPSFMTMDQSRKVLLIGKSINFLHQVCHDQTPTTKMIAVTKSAESPQDAADLFTDLENAFQGKIDAAYFETSKYLLDVLNKKYSLLDHMQAMRRYLLLGQGDFIRHLMDLLKPELVRPATTLYQHNLTGILETAVRATNAQFDSPEILRRLDVRLLEVSPGDTGWDVFSLDYHVDGPIATVFTRECMSHYLRVFNFLWRAKRMEYILTDIRKGHMCNAKLLRNMPEFSGVLHQCHILASEMVHFIHQMQYYITFEVLECSWDELWNKVQQAQDLDHIIAAHEVFLDTIISRCLLDSDSRALLNQLRAVFDQIIELQNAQDAIYRAALEELQRRLQFEEKKKQREIEGQWGVTAAEEEEENKRIGEFKESIPKMCSQLRILTHFYQGIVQQFLVLLTTSSDESLRFLSFRLDFNEHYKAREPRLRVSLGTRGRRSSHT
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline CD Markers, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10426
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.