missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBGCP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | TUBGCP3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617180
|
Novus Biologicals
NBP2-93107-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618421
|
Novus Biologicals
NBP2-93107-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TUBGCP3 Polyclonal antibody specifically detects TUBGCP3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TUBGCP3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| CD Markers, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Immunology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 10426 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Gamma-ring complex protein 104 kDa, gamma-tubulin complex component 3, GCP-3, GCP3h104p, hGCP3, hGrip104, hSpc98, SPBC98, Spc98p, Spindle pole body protein Spc98 homolog, tubulin, gamma complex associated protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human TUBGCP3 (NP_001273207.1). MATPDQKSPNVLLQNLCCRILGRSEADVAQQFQYAVRVIGSNFAPTVERDEFLVAEKIKKELIRQRREADAALFSELHRKLHSQGVLKNKWSILYLLLSLSEDPRRQPSKVSSYATLFAQALPRDAHSTPYYYARPQTLPLSYQDRSAQSAQSSGSVGSSGISSIGLCALSGPAPAPQSLLPGQSNQAPGVGDCLRQQLGSRLAWTLTANQPSSQATTSKGVPSAVSRNMTRSRREGDTGGTMEITEAAL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title