Learn More
TUBGCP2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25856-1-AP-150UL
This item is not returnable.
View return policy
Description
The TUBGCP2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TUBGCP2. This antibody recognizes human antigen. The TUBGCP2 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| TUBGCP2 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 10844 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human |