missing translation for 'onlineSavingsMsg'
Learn More

TUBGCP2 Antibody, Novus Biologicals™

Product Code. 18328319 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328319 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328319 Supplier Novus Biologicals Supplier No. H00010844D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TUBGCP2 Polyclonal antibody specifically detects TUBGCP2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBGCP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_006650.1
Gene Alias Gamma-ring complex protein 103 kDa, gamma-tubulin complex component 2, gamma-tubulin complex protein 2, GCP-2, GCP2hGrip103, h103p, hGCP2, hSpc97, MGC138162, SPBC97, Spc97p, Spindle pole body protein Spc97 homolog, tubulin, gamma complex associated protein 2
Host Species Rabbit
Immunogen TUBGCP2 (NP_006650.1, 1 a.a. - 902 a.a.) full-length human protein. MSEFRIHHDVNELLSLLRVHGGDGAEVYIDLLQKNRTPYVTTTVSAHSAKVKIAEFSRTPEDFLKKYDELKSKNTRNLDPLVYLLSKLTEDKETLQYLQQNAKERAELAAAAVGSSTTSINVPAAASKISMQELEELRKQLGSVATGSTLQQSLELKRKMLRDKQNKKNSGQHLPIFPAWVYERPALIGDFLIGAGISTDTALPIGTLPLASQESAVVEDLLYVLVGVDGRYVSAQPLAGRQSRTFLVDPNLDLSIRELVHRILPVAASYSAVTRFIEEKSSFEYGQVNHALAAAMRTLVKEHLILVSQLEQLHRQGLLSLQKLWFYIQPAMRTMDILASLATSVDKGECLGGSTLSLLHDRSFSYTGDSQAQELCLYLTKAASAPYFEVLEKWIYRGIIHDPYSEFMVEEHELRKERIQEDYNDKYWDQRYTIVQQQIPSFLQKMADKILSTGKYLNVVRECGHDVTCPVAKEIIYTLKERAYVEQIEKAFNYASKVLLDFLMEEKELVAHLRSIKRYFLMDQGDFFVHFMDLAEEELRKPVEDITPPRLEALLELALRMSTANTDPFKDDLKIDLMPHDLITQLLRVLAIETKQEKAMAHADPTELALSGLEAFSFDYIVKWPLSLIINRKALTRYQMLFRHMFYCKHVERQLCSVWISNKTAKQHSLHSAQWFAGAFTLRQRMLNFVQNIQYYMMFEVMEPTWHILEKNLKSASNIDDVLGHHTGFLDTCLKDCMLTNPELLKVFSKLMSVCVMFTNCMQKFTQSMKLDGELGGQTLEHSTVLGLPAGAEERARKELARKHLAEHADTVQLVSGFEATINKFDKNFSAHLLDLLARLSIYSTSDCEHGMASVISRLDFNGFYTERLERLSAERSQKATPQVPVLRGPPAPAPRVAVTAQ
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10844
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.