missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBGCP2 Antibody (GCP2-01) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21793
This item is not returnable.
View return policy
Description
TUBGCP2 Monoclonal specifically detects TUBGCP2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| TUBGCP2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:300, Immunocytochemistry/Immunofluorescence 1:50-1:300, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| TUBGCP2 | |
| GST-fusion protein containing amino acids 2-194 of mouse GCP2. | |
| Protein A purified | |
| RUO | |
| 10844 | |
| Human, Mouse, Rat, Pig | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| GCP2-01 | |
| Unconjugated | |
| Gamma-ring complex protein 103 kDa, gamma-tubulin complex component 2, gamma-tubulin complex protein 2, GCP-2, GCP2hGrip103, h103p, hGCP2, hSpc97, MGC138162, SPBC97, Spc97p, Spindle pole body protein Spc97 homolog, tubulin, gamma complex associated protein 2 | |
| Mouse | |
| 103 kDa | |
| 0.1 mg | |
| Primary | |
| The mouse monoclonal antibody GCP2-01 recognizes gamma-tubulin complex component 2 (GCP2), a 95-103 kDa protein. It recognizes also phosphorylated GCP2. | |
| Store at 4C. Do not freeze. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction