missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUBG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570098
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%; Zebrafish: 83%.
Tubulin is the major constituent of microtubules. The gamma chain is found at microtubule organizing centers (MTOC) such as the spindle poles or the centrosome. Pericentriolar matrix component that regulates alpha/beta chain minus-end nucleation, centrosome duplication and spindle formation.
Specifications
| TUBG2 | |
| Polyclonal | |
| Unconjugated | |
| Tubg2 | |
| AI504772; Gamma-2-tubulin; Tubg2; Tubgl; tubulin gamma 2; tubulin gamma-2 chain; tubulin, gamma 2; Unknown (protein for MGC:133882) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27175 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NRH3 | |
| Tubg2 | |
| synthetic peptide directed towards the middle region of human TUBG2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction